Orders LinkContactUsLinkComponent
VWR®, Magnetic stirring bars, triangular
VWR®, Magnetic stirring bars, triangular
35 mm, 9 mm
Catalog # 442-0388
Supplier:   VWR Collection
VWR®, Magnetic stirring bars, triangular
Catalog # 442-0388
Supplier:  VWR Collection
VWR®, Magnetic stirring bars, triangular
VWR®, Magnetic stirring bars, triangular
35 mm, 9 mm
Price
38,00 €
Log in for availability
1 * 10 items
Product Details & Documents
PTFE covered, strong Alnico V magnetic core. Smooth surface. Particularly effective for dissolving solids and mixing sediments because of the scraper-like action on the base of the container.
  • High turbulence, even at low speeds
  • Excellent chemical resistance
  • Temperature resistant, suitable for high and low temperature applications (−200 to +280 °C)
Specifications
  • Ø:
    9 mm
  • Length:
    35 mm