ContactUsLinkComponent
Model 3510 pH meter supplied with glass combination electrode (924 005), electrode holder, ATC probe (027 500), pH 4, 7 and 10 buffer sachets, European power supply, BNC shorting plug (009 146), condensed operating instructions and instruction manual 1 * 1 items
Catalog # JENW35459-06
Supplier:  Cole-Parmer
Model 3510 pH meter supplied with glass combination electrode (924 005), electrode holder, ATC probe (027 500), pH 4, 7 and 10 buffer sachets, European power supply, BNC shorting plug (009 146), condensed operating instructions and instruction manual 1 * 1 items
Catalog # JENW35459-06
Supplier:  Cole-Parmer
Frequently Bought Together