Orders LinkContactUsLinkComponent
Microscope slides, UniMark®
Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Orange
Catalog # 630-1603
Microscope slides, UniMark®
Catalog # 630-1603
Supplier:  
Microscope slides, UniMark®
Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Orange
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
  • Frosted end for labelling
  • Soda-lime glass, Hydrolytic Class 3
  • Different colours to identify preparations securely
  • Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Specifications
  • Colour:
    Orange
  • Description:
    Boxes (50) of 50 slides in an aluminium bag
  • Seq:
    12