Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Blue
Catalog # 630-1601
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Microslide blue 90 ground edges alu unimark 1 * 2.500 items Catalog #MARI0704307
Product MARI0704307 is a replacement for 630-1601 but varies by Brand, Packaging or Quantity.
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
- Frosted end for labelling
- Soda-lime glass, Hydrolytic Class 3
- Different colours to identify preparations securely
- Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Documents
Certificates
Search by Lot NumberCoC_MARI1000612_823410
Conformity declaration
MARI0704102_TDS_EN
Datasheet
Specifications
- Colour:Blue
- Description:Boxes (50) of 50 slides in an aluminium bag
- Seq:10
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.