Orders LinkContactUsLinkComponent
Microscope slides, UniMark®
Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Blue
Catalog # 630-1601
Microscope slides, UniMark®
Catalog # 630-1601
Supplier:  
Microscope slides, UniMark®
Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Blue
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Microslide blue 90 ground edges alu unimark 1 * 2.500 items Catalog #MARI0704307
Product MARI0704307 is a replacement for 630-1601 but varies by Brand, Packaging or Quantity.
Product Details & Documents
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
  • Frosted end for labelling
  • Soda-lime glass, Hydrolytic Class 3
  • Different colours to identify preparations securely
  • Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
Documents
Certificates
Search by Lot Number
Specifications
  • Colour:
    Blue
  • Description:
    Boxes (50) of 50 slides in an aluminium bag
  • Seq:
    10