Microscope slides, UniMark®
Boxes (50) of 50 slides in an aluminium bag, Yellow
630-1599
The bright printed colours of UniMark® slides frosted ends contrast well with the markings.
- Frosted end for labelling
- Soda-lime glass, Hydrolytic Class 3
- Different colours to identify preparations securely
- Thin coating to facilitate the processing of slides in automated systems
W×D×H: 76×26×1 mm
For in vitro diagnostic (IVD) applications.
- Colour:Yellow
- Description:Boxes (50) of 50 slides in an aluminium bag
- Seq:08