Tissue processing/embedding cassettes, Slimsette®, M509
Purple
720-1585
Cassette design is similar to that of the M507 model but smaller, and with a lid that does not exceed the dimensions of the cassette. Made of acetal polymer.
- Small size enables more cassettes to be loaded into labelling machines and storage systems at one time
- Highly resistant to the chemical action of histological solvents
- With 114 openings (each 1×5 mm) for efficient fluid exchange
- Integral lid
- Front face inclined 45° to enable use with certain automatic labelling systems
W×D×H: 41×28,5×6 mm
Packaging: One box with 500 cassettes.
- Colour:Purple