Orders LinkContactUsLinkComponent
Biopsy processing/embedding cassettes, Microsette™ I M502
Biopsy processing/embedding cassettes, Microsette™ I M502
Hi-vis pink
Catalog # 720-1543
Biopsy processing/embedding cassettes, Microsette™ I M502
Catalog # 720-1543
Supplier:  
Biopsy processing/embedding cassettes, Microsette™ I M502
Biopsy processing/embedding cassettes, Microsette™ I M502
Hi-vis pink
Discontinued
Product Details & Documents
Acetal polymer.
  • No need for biopsy pads
  • Large compartment 25×30 mm
  • 2000 holes (Ø 0,26 mm) suitable for needle biopsies
  • Integral lid
  • Anterior writing area 45° angle suitable for some labelling systems
Specifications
  • Colour:
    Hi-vis pink