Staining systems for microscope slides, DrainRack Junior
Staining systems, Blue, 100
631-1915
This rugged tray used as a drain rack can hold up to 100 microscope slides in 50 individually numbered slots. All slides are stored upright for easier insertion and removal.
- Barcodes can be read without having to remove the slides from the box
- To save space, the number of slides in each slot can be doubled simply by inserting two slides in each slot
- Not autoclavable
- No. of slides:100
- Colour:Blue
- Description:Staining systems