Slide boxes, SlideFile™ Junior
White, PS, 200, 82×140×86 mm
Catalog # 631-1908
Supplier: Simport Scientific
Jump to:
Product Details & Documents
Boxes are impact-resistant and stackable for space efficient storage.
- Barcodes can be read through the clear lid
- Number of slides can be doubled by storing two in a slot
- Tinted hinged cover makes the content easy to see
Ordering information: Include a slide box and a removable tray.
Specifications
- No. of slides:200
- Colour:White
- Material:PS
- W×D×H:82×140×86 mm