Orders LinkContactUsLinkComponent
Slide holders, SlideFolder™
Slide holders, SlideFolder™
Slide holders, Opaque/white, Plastic, 20
Catalog # 630-1028
Supplier:   Simport Scientific
Slide holders, SlideFolder™
Catalog # 630-1028
Supplier:  Simport Scientific
Slide holders, SlideFolder™
Slide holders, Opaque/white, Plastic, 20
Price
294,00 €
Log in for availability
1 * 10 items
Product Details & Documents
The SlideFolder is manufactured from heat-resistant, injection moulded plastic and can hold up to 20 standard microscope slides.
  • Numbered spaces and transparent leaves for easier identification
  • Five colours allowing colour coded classification of the slides
  • Slides are removed simply by pressing on one end
  • Can be stacked to save space
Specifications
  • No. of slides:
    20
  • Colour:
    Opaque/white
  • Description:
    Slide holders
  • Material:
    Plastic