Orders LinkContactUsLinkComponent
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
10 mm, 13 mm
Catalog # 229-0232
Supplier:   SciLabware
Stopcocks, Rotaflo®
Catalog # 229-0232
Supplier:  SciLabware
Stopcocks, Rotaflo®
Stopcocks, Rotaflo®
10 mm, 13 mm
Price
188,00 €
Log in for availability
1 * 1 items
Product Details & Documents
PYREX®, borosilicate glass with PTFE needle valve.
  • Lubricant is not required (no seizure, no contamination with grease)
  • Interchangeable elements
  • Precise flow control
These chemically inert stopcocks can be used at temperatures up to +200 °C. Two different models are available: general purpose (vacuum rating up to 10-4 mm Hg) and high performance (vacuum rating up to 10-9 mm Hg).
Specifications
  • Hole Ø:
    10 mm
  • Seq:
    18
  • For tube I-Ø:
    13 mm
Accessories
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 6 mm
    Replacement key, bore Ø 6 mm
    229-0230
    High performance Rotaflo® stopcocks
    Log in for availability
    1 * 2 items - 113,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 6 mm
    Replacement key, bore Ø 6 mm
    229-0231
    General purpose Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 156,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement locking ring for key with bore 6 mm
    Replacement locking ring for key with bore 6 mm
    BIBBLR6
    Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 37,20 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 10 mm
    Replacement key, bore Ø 10 mm
    229-0226
    High performance Rotaflo® stopcocks
    Log in for availability
    1 * 1 items - 72,30 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 3 mm
    Replacement key, bore Ø 3 mm
    229-0229
    High performance Rotaflo® stopcocks
    Log in for availability
    1 * 2 items - 80,60 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement locking ring for key with bore 3 mm
    Replacement locking ring for key with bore 3 mm
    BIBBLR3
    Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 25,40 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 10 mm
    Replacement key, bore Ø 10 mm
    229-0234
    General purpose Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 242,00 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement key, bore Ø 3 mm
    Replacement key, bore Ø 3 mm
    BIBBGP3K
    General purpose Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 99,40 €
Product Information
  • Catalog #
    For
    Availability
    Price
  • Replacement locking ring for key with bore 10 mm
    Replacement locking ring for key with bore 10 mm
    BIBBLR10
    Rotaflo® stopcocks
    Log in for availability
    1 * 5 items - 35,40 €