Orders LinkContactUsLinkComponent
Microscope slides, SuperFrost®, Menzel Gläser
Orange, Unwrapped
Catalog # 630-2858
Microscope slides, SuperFrost®, Menzel Gläser
Catalog # 630-2858
Supplier:  
Microscope slides, SuperFrost®, Menzel Gläser
Microscope slides, SuperFrost®, Menzel Gläser
Orange, Unwrapped
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
These slides are made of extra-white glass, washed clean and polished.
  • Labelling area resistant to all common solvents
  • Six colours for identification and colour coding
  • Thick labelling area to prevent slides from sticking together or getting scratched
Suitable for in vitro diagnostic (IVD) applications.
Complies with ISO 8037/I
 : ISO 8037/I.
 : In cardboard boxes of 50.
Specifications
  • Packed:
    Unwrapped
  • Colour:
    Orange
  • Seq:
    223