Orders LinkContactUsLinkComponent
Capillary tubes
Capillary tubes
Both ends open, 75 mm, 1,8/2,0 mm
Catalog # 612-4866
Supplier:   MURRAY S
Capillary tubes
Catalog # 612-4866
Supplier:  MURRAY S
Capillary tubes
Capillary tubes
Both ends open, 75 mm, 1,8/2,0 mm
Price
125,00 €
Log in for availability
1 * 1.000 items
Product Details & Documents
Soda glass.
  • For melting point determinations
Specifications
  • Ext.Ø:
    1,8/2,0 mm
  • Description:
    Both ends open
  • Length:
    75 mm