Orders LinkContactUsLinkComponent
Group Lock Boxes, Portable
Group Lock Boxes, Portable
Group lock box, mild sheet steel, for approx. 40 padlocks, Blue, 227×89×152 mm
Catalog # BRDY045190
Supplier:   Brady
Group Lock Boxes, Portable
Catalog # BRDY045190
Supplier:  Brady
Group Lock Boxes, Portable
Group Lock Boxes, Portable
Group lock box, mild sheet steel, for approx. 40 padlocks, Blue, 227×89×152 mm
Price
86,00 €
Log in for availability
1 * 1 items
Product Details & Documents
For group lockout situations involving a large number of workers and equipment, lock boxes can offer a number of advantages over hasps.
  • Made of mild sheet steel
  • Slot in front allows keys to be inserted when box is locked
Can also be used as a lock storage box holding up to 40 padlocks.
Specifications
  • Colour:
    Blue
  • Description:
    Group lock box, mild sheet steel, for approx. 40 padlocks
  • Seq:
    01
  • W×D×H:
    227×89×152 mm