First-aid kits
Medium first aid kit, 11 - 20 people
Catalog # 118-0122
Supplier: SAFETY FIRST AID GROUP
Jump to:
Product Details & Documents
The contents of the following first aid kits meet the requirements as specified in the HSE Approved Code of Practice. The code of practice states 'An employer should make an assessment of the first aid needs appropriate to the circumstance of each workplace'. To assist in selecting the appropriate kit, employee ratios are given as guidance provided by the British Healthcare Trade Association.
Delivery information: All kits are supplied with a first aid guidance leaflet.
Specifications
- Description:Medium first aid kit, 11 - 20 people
- Cat. No.:118-0122
- Contents:40 × Waterproof plasters, sterile
10 × individually wrapped wipes
4 × eye pads with bandages, non woven
6 × triangular bandages, non woven
12 × Safety pins
9 × sterile dressing, medium size
3 × sterile dressing, large size
2 pairs of disposable examination gloves
1 wall bracket - Model:Standard (11 - 20 persons)