Micro pipettes, ringcaps®
50/100 µl, 125 mm, Standard
Catalog # 612-2410
Supplier: HIRSCHMANN
Jump to:
Product Details & Documents
DURAN®, borosilicate glass, with ring mark, disposable.
- Colour coded according to ISO for clear identification
- Conformity certified
ISO 9000, DIN ISO 7550, IVD 98/79 EC
: ISO 9000, DIN ISO 7550, IVD 98/79 EC
Specifications
- Type:Standard
- Capacity:50/100 µl
- Length:125 mm