Orders LinkContactUsLinkComponent
Test tubes without rim, medium wall
Test tubes without rim, medium wall
150 mm, 24 mm
Catalog # 212-0462
Supplier:   MURRAY S
Test tubes without rim, medium wall
Catalog # 212-0462
Supplier:  MURRAY S
Test tubes without rim, medium wall
Test tubes without rim, medium wall
150 mm, 24 mm
Price
80,40 €
Log in for availability
1 * 50 items
Product Details & Documents
Borosilicate glass, grade 3.3, SAMCO '33'. Medium walled.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Suitable for all usual laboratory applications. They will withstand temperature change up to and including boiling of samples.
Certifications: ISO 4142:2002
Specifications
  • Ext.Ø:
    24 mm
  • Length:
    150 mm