Orders LinkContactUsLinkComponent
Charts
Charts
C472, –20...+50 °C; 0 - 100 % RH, 24 hours
Catalog # 620-0413
Supplier:   DICKSON
Charts
Catalog # 620-0413
Supplier:  DICKSON
Charts
C472, –20...+50 °C; 0 - 100 % RH, 24 hours
Price
79,10 €
Log in for availability
1 * 60 items
Product Details & Documents
Specifications
  • Description:
    C472
  • Seq:
    12
  • Range:
    –20...+50 °C; 0 - 100 % RH
  • Recording time:
    24 hours