Cavity slides
Cavity slides with epoxy coating, Black, 21, 4 mm
Catalog # 631-0460
Supplier: Epredia
Jump to:
Product Details & Documents
These autoclavable slides are specially designed for immunofluorescence.
- Adhesive surface and hydrophobic mask
- Hydrophobic epoxy- or PTFE-coating
- (*) with rings or wells
Developped for clinical diagnosis applications.
: Available with rings or wells (*).
Documents
Specifications
- No. of wells:21
- Colour:Black
- Description:Cavity slides with epoxy coating
- Seq:013
- Well Ø:4 mm