Microtubes, with Thread
Micro tubes with neutral screw cap assembled, sterile, skirted base, 0,5 ml, 1000/case
Catalog # SARS72.730.005
Supplier: SARSTEDT
Jump to:
Product Details & Documents
PP, translucent.
- Safe screw cap with O-ring
- Sterile or non sterile
- Autoclavable
For transport, storage and sample processing in the laboratory.
Documents
Specifications
- Packed:1000/case
- Capacity:0,5 ml
- Description:Micro tubes with neutral screw cap assembled, sterile, skirted base
- Seq:2