Orders LinkContactUsLinkComponent
Sample bags, Duratec
Sample bags, Duratec
5 l, 305×305 mm
Catalog # VARICP12222
Supplier:   VARIAN
Sample bags, Duratec
Catalog # VARICP12222
Supplier:  VARIAN
Sample bags, Duratec
Sample bags, Duratec
5 l, 305×305 mm
Price
678,25 €
Log in for availability
1 * 10 items
Product Details & Documents
Specifications
  • Capacity:
    5 l
  • L×W:
    305×305 mm
  • Seq:
    03