Adhesion slides
Adhesion slides with 3 wells (Ø = 15 mm)
Catalog # MARI0901100
Supplier: MARIENFELD
Jump to:
Product Details & Documents
Adhesion slides with frosted ends and 90° ground edges.
Specifications
- Description:Adhesion slides with 3 wells (Ø = 15 mm)