Order Entry
Slovenia
Orders LinkContactUsLinkComponent
 

Volumetric flasks, class A, blue markings

20 ml, 12/21, 105 ± 5 mm, 0,040 ± ml
  HIRS2820366
 :   HIRSCHMANN
Volumetric flasks, class A, blue markings
  HIRS2820366
Volumetric flasks, class A, blue markings
Volumetric flasks, class A, blue markings

20 ml, 12/21, 105 ± 5 mm, 0,040 ± ml

 
40,50 €
   
1 * 2 items
 

DURAN®, borosilicate glass, without stopper, conformity certified (DE-M marking).

  • Blue enamelled indelible markings resistant to acids and alkalis
  • Certificate of conformity with dated batch identification available via website: (www.hirschmannlab.de)

*Wide neck

 : DIN EN ISO 1042

 
  • NS:
    12/21
  • Capacity:
    20 ml
  • Height:
    105 ± 5 mm
  • Seq:
    11
  • Tolerance:
    0,040 ± ml