Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
500 ml, 19/26, 270 mm, 0,25 ± ml
Catalog # HIRS7570190
Supplier:   HIRSCHMANN
Volumetric flasks, class A
Catalog # HIRS7570190
Supplier:  HIRSCHMANN
Volumetric flasks, class A
Volumetric flasks, class A
500 ml, 19/26, 270 mm, 0,25 ± ml
Price
64,70 €
Log in for availability
1 * 2 items
Product Details & Documents
PMP, translucent, with PP NS-stopper.
Specifications
  • NS:
    19/26
  • Capacity:
    500 ml
  • Height:
    270 mm
  • Tolerance:
    0,25 ± ml