Orders LinkContactUsLinkComponent
Volumetric flasks, class A
Volumetric flasks, class A
50 ml, 10/19, 150 mm, 0,06 ± ml
Catalog # HIRS7570175
Supplier:   HIRSCHMANN
Volumetric flasks, class A
Catalog # HIRS7570175
Supplier:  HIRSCHMANN
Volumetric flasks, class A
Volumetric flasks, class A
50 ml, 10/19, 150 mm, 0,06 ± ml
Price
43,60 €
Log in for availability
1 * 2 items
Product Details & Documents
PMP, translucent, with PP NS-stopper.
Specifications
  • NS:
    10/19
  • Capacity:
    50 ml
  • Height:
    150 mm
  • Tolerance:
    0,06 ± ml