Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
N1-(2,6-dichloro-3-pyridyl)-2-{[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-3-yl]sulfanyl}acetamide, TEC H 1 * 250 mg
Catalog # MAYBSPB07891.250
Supplier:
Thermo Scientific
Expand image
N1-(2,6-dichloro-3-pyridyl)-2-{[4-(2,3-dihydro-1,4-benzodioxin-6-yl)-5-oxo-4,5-dihydro-1H-1,2,4-triazol-3-yl]sulfanyl}acetamide, TEC H 1 * 250 mg
Catalog # MAYBSPB07891.250
Supplier:
Thermo Scientific
Frequently Bought Together
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Sealing film for manual application, Parafilm® M
Supplier: HEATHROW
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Water, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
VWR® Facial Tissues
Supplier: VWR Collection
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Transfer Pipettes, High Performance
Supplier: VWR Collection
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® Serological Pipette, Standard Line
Supplier: VWR Collection