2,3,4,5-Tetrachlorophenol
Glass bottle for solids, 1 g
Catalog # MAYBBTB15035.1
Supplier: Thermo Scientific
Jump to:
Product Details & Documents
- CAS Number:4901-51-3
- Formula:C₆H₂Cl₄O
- Molecular Weight:231.89 g/mol
- Boiling Pt:289 °C (1013 hPa)
- Melting Pt:116 °C
- Density:1.6 g/cm³ (20 °C)
- EINECS:225-531-7
- UN:2020
- ADR:6.1,III
- :Density: 1.6 g/cm³ (20 °C)
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g



