Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Slovenia
Order Entry
Order Status
Your Account
Cart
Orders Link
ContactUsLinkComponent
Home
Forceps
Forceps, sharp
Print
Forceps, sharp
Sharp/bent, 200 mm
Catalog # 232-0116
Supplier:
BOCHEM
Forceps, sharp
Catalog # 232-0116
Supplier:
BOCHEM
Return to Top
Forceps, sharp
Sharp/bent, 200 mm
Specifications
Jump to:
Specifications
Price
8,00 €
Log in
for availability
1 * 1 items
-
+
Other Product Family Options Available
Return Policy
Product Details & Documents
Stainless steel ¹⁸/₁₀, antimagnetic, bent.
Specifications
Type:
Sharp/bent
Length:
200 mm
Silica gel, granules, Chameleon® C 2 - 6 mm drying agent
#: 83000.290
Supplier: VWR Chemicals