Single-component explosives solutions
Internal standard includes 4-amino-2,6-dinitrotoluene, with 1000 µg/ml in acetonitrile, 1 ml/ampule, 1
REST31671
: Restek
Single-component explosives solutions supports US department of defense base closures and remediation.
- Mixtures and singles supporting LC US EPA method 8330
- Mixtures and singles supporting GC-ECD US EPA method 8095
- Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.
- Pk:1
- Description:Internal standard includes 4-amino-2,6-dinitrotoluene, with 1000 µg/ml in acetonitrile, 1 ml/ampule