Orders LinkContactUsLinkComponent
Single-component explosives solutions
Single-component explosives solutions
Internal standard includes nitrobenzene, with 1000 µg/ml in acetonitrile, 1 ml/ampule, 1
Catalog # REST31657
Supplier:   Restek
Single-component explosives solutions
Catalog # REST31657
Supplier:  Restek
CAS Number:  
Single-component explosives solutions
Single-component explosives solutions
Internal standard includes nitrobenzene, with 1000 µg/ml in acetonitrile, 1 ml/ampule, 1
CAS Number:  
Price
50,50 €
Log in for availability
1 * 1 items
Product Details & Documents
Single-component explosives solutions supports US department of defense base closures and remediation.
  • Mixtures and singles supporting LC US EPA method 8330
  • Mixtures and singles supporting GC-ECD US EPA method 8095
  • Internal standards and surrogates to support both methods
These materials support nitroaromatic, nitramine, and nitroester analyses by GC-ECD (Method 8095). Compounds listed are explosives, manufacturing intermediates, or degradation products. Method 8095 mixtures contains the components at concentration ratios appropriate for ECD.
Specifications
Specifications
  • Pk:
    1
  • Description:
    Internal standard includes nitrobenzene, with 1000 µg/ml in acetonitrile, 1 ml/ampule