PCB congeners
PCB congener standard includes 2,3',4,4',5-pentachlorobiphenyl (BZ #118) with 10 µg/ml in isooctane, 1 ml/ampoule, 1 mL
Catalog # REST32293
Supplier: Restek
Jump to:
Product Details & Documents
Certified reference materials.
Manufactured and QC-tested in ISO-accredited labs.
Specifications
Specifications
- Pk:1 mL
- Description:PCB congener standard includes 2,3',4,4',5-pentachlorobiphenyl (BZ #118) with 10 µg/ml in isooctane, 1 ml/ampoule
- Cat. No.:REST32293
- Formula:C<sub>12</sub>H<sub>5</sub>Cl<sub>5</sub>
- Description:2,3',4,4',5-pentachlorobiphenyl (BZ #118)
- CAS No.:31508-00-6
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



