PCB congeners
PCB congener standard includes 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153) with 10 µg/ml in isooctane, 1 ml/ampoule, 1
REST32287
: Restek
Certified reference materials.
Manufactured and QC-tested in ISO-accredited labs.
- Pk:1
- Description:PCB congener standard includes 2,2',4,4',5,5'-hexachlorobiphenyl (BZ #153) with 10 µg/ml in isooctane, 1 ml/ampoule
- Cat. No.:REST32287
- Formula:C<sub>12</sub>H<sub>4</sub>Cl<sub>6</sub>
- Description:2,2',4,4',5,5'-hexachlorobiphenyl<br /> (BZ #153)
- CAS No.:35065-27-1



