Order Entry
Slovenia
Orders LinkContactUsLinkComponent
PCB congeners

PCB congeners

PCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampoule, 1
Catalog # REST32284
Supplier:   Restek
PCB congeners
Catalog # REST32284
Supplier:  Restek
CAS Number:  
PCB congeners
PCB congeners

PCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampoule, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 items
Product Details & Documents

Certified reference materials.

Manufactured and QC-tested in ISO-accredited labs.

Specifications

Specifications

  • Pk:
    1
  • Description:
    PCB congener standard includes2,2',5,5'-tetrachlorobiphenyl (BZ #52) with 10 µg/ml in isooctane, 1 ml/ampoule
  • Cat. No.:
    REST32284
  • Formula:
    C<sub>12</sub>H<sub>6</sub>Cl<sub>4</sub>
  • Description:
    2,2',5,5'-tetrachlorobiphenyl (BZ #52)
  • CAS No.:
    35693-99-3

Specifications