Automatic viscometers, ViscoSystem® AVS® 470
The ViscoSystem AVS 470 is the first measuring device that allows suction and pressure measurements completely independent of a PC.
- Determination with pressure or with vacuum for high viscosities
- Both detections methods optical or thermal are possible
- Nearly all necessary calculations are integrated
- Additional parameters can be set up over a regular PS 2 keyboard
This brings maximum independence and flexibility, allowing you to set up a measuring station, that meets highest requirements, even under difficult conditions, to monitor production or control quality in the polymer and mineral oil industry.
The ViscoSystem AVS 470 determines the time which the sample to be examined requires to flow through the measuring distance in the capillary viscometer with quartz precision.
All common types of viscosity calculations are already integrated into the device, a small PS 2 keyboard is all you need to enter additional data.
A serial printer can be used to conveniently document your measuring results.
- Model:AVS 470
- Range:0,35 − 1800 (pressure) ; 0,35 − 5000 (suction) mm²/s (cSt)
- Description:Automatic viscometer with ViscoPump II module for optoelectronic sensing
- Interfaces:RS232C for serial printer
- Cat. No.:616-0026
- Measurement range (flow time):<9999,99 s
- Temperature regulation period:0 – 20 minutes
- W×D×H:225×320×205 mm
- Viscosity range:Pressure: 0,35 - 1800 mm²/s (cSt)
Suction capacity: 0,35 - ~5000 mm²/s (cSt) - Pump pressure:160 mbar
- Time measurement accuracy:±0,01 %
- Weight:5,4 kg
- Display:LCD
- Preselection of the number of measurements:Up to 99
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- SCOI285414031VZ 5505 included with AVS®/S and AVS®/SK, to connect viscometers of type: 517 .., 520 .., 530 .., 532 .., 537 .., 538 .., 547 .., 548 ..and 549 .. to a AVS® 310, 350, 450 and 4701 * 1 items - 343,00 €
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description