Orders LinkContactUsLinkComponent
Hydrochloric acid 35%, TECHNICAL
Hydrochloric acid 35%, TECHNICAL
Plastic drum, 25 L
Catalog # 20246.460
Corrosive
Irritant
Supplier:   VWR Chemicals
Hydrochloric acid 35%, TECHNICAL
Catalog # 20246.460
Supplier:  VWR Chemicals
CAS Number:  7647-01-0
Hydrochloric acid 35%, TECHNICAL
Hydrochloric acid 35%, TECHNICAL
Plastic drum, 25 L
CAS Number:  7647-01-0
Price
139,00 €
Log in for availability
1 * 25 L
Product Details & Documents
Synonyms:  Hydrochloric acid solution, Muriatic acid
  • CAS Number:7647-01-0
  • Formula:HCl
  • MDL Number:MFCD00011324
  • Molecular Weight:36.46 g/mol
  • Boiling Pt:110 °C (1013 hPa)
  • Melting Pt:–30 °C
  • Density:1.18 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-595-7
  • UN:1789
  • ADR:8,II
  • REACH:01-2119484862-27
Specifications
Specification Test Results
  • Assay:
    Min. 35 %
Specifications
  • Pack type:
    Plastic drum
  • Pk:
    25 L