Orders LinkContactUsLinkComponent
Section lifters
Section lifters
150 mm, 50 mm, 20 mm
Catalog # RSGA063.020
Supplier:   RSG
Section lifters
Catalog # RSGA063.020
Supplier:  RSG
Section lifters
Section lifters
150 mm, 50 mm, 20 mm
Price
60,80 €
Log in for availability
1 * 5 items
Product Details & Documents
Blade made of stainless steel, wooden handle.
  • Blade with excellent heat resistance
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    150 mm
  • Blade width:
    20 mm
  • Blade length:
    50 mm