Orders LinkContactUsLinkComponent
Spoon spatulas, micro
Spoon spatulas, micro
185 mm, 5 mm
Catalog # 231-0191
Supplier:   RSG
Spoon spatulas, micro
Catalog # 231-0191
Supplier:  RSG
Spoon spatulas, micro
Spoon spatulas, micro
185 mm, 5 mm
Price
43,70 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic.
  • With a micro spoon at one end and a micro spatula at the other
  • Excellent chemical resistance
  • Excellent thermal resistance
  • Quick and easy to clean
Specifications
  • Length:
    185 mm
  • Blade width:
    5 mm