Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
185 mm, 3 mm
Catalog # 231-0462
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0462
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro
185 mm, 3 mm
Price
36,80 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatulas with round handle, one flat and one rounded end.
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Very flexible blades
Documents
Specifications
  • Length:
    185 mm
  • Blade width:
    3 mm