Spoon spatulas, flat
Single spoon spatula, flat ending, 180 mm, 10 mm
RSGA098.180
: RSG
Flat spoon spatulas made of stainless steel. Available singe or double ended.
- Description:Single spoon spatula, flat ending
- Length:180 mm
- Blade width:10 mm