Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
150 mm, 7 mm
Catalog # 231-0399
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0399
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro
150 mm, 7 mm
Price
59,10 €
Log in for availability
1 * 10 items
Product Details & Documents
Stainless steel 18/10, anti-magnetic. Multipurpose spatula for a wide range of uses.
  • Both ends have an angled spoon
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
  • Quick and easy to clean
Specifications
  • Length:
    150 mm
  • Blade width:
    7 mm