Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff
Double spatulas, micro, stiff
185 mm, 3 mm
Catalog # RSGA833.185
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA833.185
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff
185 mm, 3 mm
Price
28,00 €
Log in for availability
1 * 5 items
Product Details & Documents
Rigid micro double spatulas made of stainless steel.
Specifications
  • Length:
    185 mm
  • Blade width:
    3 mm