Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff
Double spatulas, micro, stiff
150 mm, 5 mm
Catalog # RSGA835.150
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA835.150
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff
150 mm, 5 mm
Price
27,50 €
Log in for availability
1 * 5 items
Product Details & Documents
Rigid micro double spatulas made of stainless steel.
Specifications
  • Length:
    150 mm
  • Blade width:
    5 mm