Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff
Double spatulas, micro, stiff
130 mm, 6 mm
Catalog # RSGA836.130
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA836.130
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff
130 mm, 6 mm
Price
27,10 €
Log in for availability
1 * 5 items
Product Details & Documents
Rigid micro double spatulas made of stainless steel.
Specifications
  • Length:
    130 mm
  • Blade width:
    6 mm