Orders LinkContactUsLinkComponent
Double spatulas, micro, stiff
Double spatulas, micro, stiff
150 mm, 6 mm
Catalog # RSGA836.150
Supplier:   RSG
Double spatulas, micro, stiff
Catalog # RSGA836.150
Supplier:  RSG
Double spatulas, micro, stiff
Double spatulas, micro, stiff
150 mm, 6 mm
Price
27,60 €
Log in for availability
1 * 5 items
Product Details & Documents
Rigid micro double spatulas made of stainless steel.
Specifications
  • Length:
    150 mm
  • Blade width:
    6 mm