Orders LinkContactUsLinkComponent
Forceps, premium quality
Forceps, premium quality, Curved, pointed, 115 mm
Catalog # RSGA014.115
Supplier:   RSG
Forceps, premium quality
Catalog # RSGA014.115
Supplier:  RSG
Forceps, premium quality
Forceps, premium quality
Forceps, premium quality, Curved, pointed, 115 mm
Price
56,20 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel 1,4301.
Specifications
  • Type:
    Curved, pointed
  • Description:
    Forceps, premium quality
  • Length:
    115 mm