Orders LinkContactUsLinkComponent
Inoculating loops, reusable
Inoculating loops, reusable
Loops, length 75 mm, Ø 1,5 mm
Catalog # 631-0084
Supplier:   RSG
Inoculating loops, reusable
Catalog # 631-0084
Supplier:  RSG
Inoculating loops, reusable
Inoculating loops, reusable
Loops, length 75 mm, Ø 1,5 mm
Price
17,70 €
Log in for availability
1 * 10 items
Product Details & Documents
These NiCr 8020 loops are available in a range of sizes and lengths with internal diameter of 1,5, 2,5 or 4 mm. They are heat resistant and can be flame sterilised. Diameter of wire: 0,6 mm. Loop holder available separately.
Documents
Specifications
  • Description:
    Loops, length 75 mm, Ø 1,5 mm
Accessories
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Needle holder
    Needle holder
    391-0374
    240 mm
    Log in for availability
    1 * 1 items - 17,30 €
Product Information
  • Catalog #
    Length
    Availability
    Price
  • Needle holder
    Needle holder
    391-0373
    170 mm
    Log in for availability
    1 * 1 items - 15,00 €