Orders LinkContactUsLinkComponent
Double spatulas, micro
Double spatulas, micro
150 mm, 7 mm
Catalog # 231-0430
Supplier:   RSG
Double spatulas, micro
Catalog # 231-0430
Supplier:  RSG
Double spatulas, micro
Double spatulas, micro
150 mm, 7 mm
Price
43,20 €
Log in for availability
1 * 5 items
Product Details & Documents
Stainless steel. Anti-magnetic, very flexible micro powder spatula.
  • With round handle, one flat and one scoop shaped, rounded spatula end
  • Straight shape
  • Excellent thermal resistance, autoclavable
  • Good chemical resistance
Documents
Specifications
  • Length:
    150 mm
  • Blade width:
    7 mm