Orders LinkContactUsLinkComponent
Sample vessel
Sample vessel
Catalog # HACHLZQ035_U
Sample vessel
Catalog # HACHLZQ035_U
Supplier:  
Sample vessel
Sample vessel
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Sample vessel made from PS. For aqueous solutions, acids and bases.
For use with autosampler LPV122/LZQ031.