TLC polyester sheets
POLYGRAM® TLC polyester sheets SIL G/UV254, Silica gel, 40×80 mm
Catalog # 552-0277
Supplier: MACHEREY-NAGEL
Jump to:
Product Details & Documents
Documents
Thin Layer Chromatography.pdf
Additional Product Details
Specifications
- Coating material:Silica gel
- Plate size:40×80 mm
- Description:POLYGRAM® TLC polyester sheets SIL G/UV254
- Fluorescent indicator:F254