Orders LinkContactUsLinkComponent
Ethidium bromide solution 0,07 %
Ethidium bromide solution 0,07 %
Dropping bottle, 5 mL
Catalog # A2273.0005
Supplier:   PanReac AppliChem
Ethidium bromide solution 0,07 %
Catalog # A2273.0005
Supplier:  PanReac AppliChem
CAS Number:  1239-45-8
Ethidium bromide solution 0,07 %
Dropping bottle, 5 mL
CAS Number:  1239-45-8
Price
38,80 €
Log in for availability
1 * 5 mL
Product Details & Documents
Ethidium bromide solutions with a concentration of 0,7 mg/ml.
Delivery information: Supplied in a dropper bottle.
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Storage Temperature:Ambient
Specifications
Specifications
  • Pack type:
    Dropping bottle
  • Pk:
    5 mL